Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK 0660

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective PPARδ antagonist (IC50 values are 0.155, > 10 and ≥ 10 μM at PPARδ, PPARα and PPARγ respectively). Exhibits inverse agonist effects when administered by itself.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 418.49 |
| Purity | >99% |
| Molecular Weight (g/mol) | 418.48 |
| Chemical Name or Material | GSK 0660 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction