Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSTA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $482.00
Specifications
Antigen | GSTA3 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119147
|
Invitrogen™
PA5119147 |
200 μL |
Each for $482.00
|
|
|||||
PIPA5121942
|
Invitrogen™
PA5121942 |
20 μL |
Each for $178.40
|
|
|||||
Description
GSTA3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-tranferase belonging to the alpha class genes that are located in a cluster mapped to chromosome 6. Genes of the alpha class are highly related and encode enzymes with glutathione peroxidase activity. However, during evolution, this alpha class gene diverged accumulating mutations in the active site that resulted in differences in substrate specificity and catalytic activity. The enzyme encoded by this gene catalyzes the double bond isomerization of precursors for progesterone and testosterone during the biosynthesis of steroid hormones. An additional transcript variant has been identified, but its full length sequence has not been determined.Specifications
GSTA3 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
Gsta3 | |
glutathione S-alkyltransferase A3; glutathione S-aralkyltransferase A3; glutathione S-aryltransferase A3; glutathione S-transferase A3; glutathione S-transferase A3 subunit; glutathione S-transferase A3-3; glutathione S-transferase A5; glutathione S-transferase alpha 1; glutathione S-transferase alpha 3; glutathione S-transferase alpha-3; Glutathione S-transferase alpha-5; glutathione S-transferase Ya3; Glutathione S-transferase Yc; glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; Glutathione S-transferase Yc-2; glutathione S-transferase Yc2 subunit; glutathione S-transferase, alpha 3; Glutathione-S-transferase alpha type (Ya); GST 2-2; GST A3-3; GST A5-5; GST AA; GST class-alpha member 3; GST Yc1; GST Yc2; Gst2-3; Gsta1; GSTA3; GSTA3-3; Gsta5; Gstyc; Gstyc1; Gstyc2; GTA3; MGC22232; S-(hydroxyalkyl)glutathione lyase A3; Yc1; Yc2 | |
Gsta3 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P30115 | |
14859 | |
Recombinant protein Glutathione S Transferase Alpha 3. The antigen corresponds to amino acid range 1-221 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title