Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 7647
GREENER_CHOICE

Catalog No. p-4783503
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective, potent PPARα agonist. Orally active

Potent and highly selective PPARα agonist (EC50 values are 6, 1100 and 6200 nM for human PPARα, PPARγ and PPARδ receptors respectively). Modulates oleate metabolism and mitochondrial enzyme gene expression in mature myotubules in vitro. Has lipid-lowering effects following oral administration in vivo. Reduces NO production in macrophages; exhibits anti-inflammatory properties.

Chemical Identifiers

CAS 265129-71-3
Molecular Formula C29H46N2O3S

Specifications

Quantity 10 mg
Formula Weight 502.75
Purity >99%
Molecular Weight (g/mol) 502.76
Chemical Name or Material GW 7647
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.