Learn More
Eisco™ Gypsum Mineral Specimen
SCI_ED
Catalog No. s24689 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Gypsum Mineral Specimen ideal for use in study of cleavage and fracture as example of imperfect cleavage planes.
  • For use in learning about mineral hardness
  • Gypsum is soft enough to be scratched by fingernail
  • One of the standards on Mohs hardness scale (value of 2)
  • Similar appearance to other minerals therefor useful in identification exercises
  • Crystals grow in soft, wide sheets, but is also commonly found filling veins and fractures in other rocks
Specifications
Product Type Mineral, Gypsum
For Use With (Application) Study of cleavage and fracture as example of imperfect cleavage planes
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.