Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA95/AKAP8L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $728.30
Specifications
| Antigen | HA95/AKAP8L |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
HA95/AKAP8L Polyclonal specifically detects HA95/AKAP8L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HA95/AKAP8L | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| A kinase (PRKA) anchor protein 8-like, AKAP8-like protein, HA95, HAP95 DKFZp434L0650, helicase A-binding protein 95 kDa, Homologous to AKAP95 protein, HRIHFB2018, NAKAP, NAKAP95 A-kinase anchor protein 8-like, neighbor of A kinase anchoring protein 95, Neighbor of AKAP95, Neighbor of A-kinase-anchoring protein 95 | |
| AKAP8L | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 26993 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FLQEYVTNKTKKTEELRKTVEDLDGLIQQIYRDQDLTQEIAMEHFVKKVEAAHCAACDLFIPMQFGIIQKHLKTMDHNRNRR | |
| Primary | |
| Specificity of HA95/AKAP8L antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title