Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Shock Protein 90α/hsp90α (HSP86) Ab-1 Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Specifications

Antigen Heat Shock Protein 90alpha/hsp90alpha (HSP86) Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 7
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB119P0
View Documents
Epredia
RB119P0
Concentrated 100 μL N/A N/A N/A
RB119P1
View Documents
Epredia
RB119P1
Concentrated 500 μL N/A N/A N/A
RB119P1ABX
View Documents
Epredia
RB119P1ABX
Concentrated 500 μL N/A N/A N/A
RB119P
View Documents
Epredia
RB119P
Concentrated 1 mL N/A N/A N/A
RB119PABX
View Documents
Epredia
RB119PABX
Concentrated 1 mL N/A N/A N/A
RB119R1
View Documents
Epredia
RB119R1
Prediluted 1 mL N/A N/A N/A
RB119R7
View Documents
Epredia
RB119R7
Prediluted 7 mL N/A N/A N/A
Description

Description

Cells respond to heat and chemical stress by inducing the synthesis of a group of proteins referred to as heat shock proteins. HSP90 is present in cells at high levels in the cytoplasm and is composed of two separate gene products, HSP90α and HSP90Β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 2-12

Immunogen: A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90α (hsp90α)

Molecular Weight: 86 kDa

Positive Control: LS174T cells, MAD109 cells or breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Heat Shock Protein 90alpha/hsp90alpha (HSP86) Ab-1
Polyclonal
Rabbit
Stress Response
A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90alpha (hsp90alpha)
Primary
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Human, Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.