Learn More
Heidolph™ Platform Shaker Accessories - Clamps
SDP
For use with Perforated Platform 100, 1000, and 2000 when used in conjunction with Heidolph platform shakers
$63.86 - $186.43
Specifications
Product Type Flask Clamp
View More Specs
Products 7
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
13-900-838
SDP
View Documents
Heidolph™
036303390
25 mL Erlenmeyer Flask
Each for $63.86
Only null left
Add to Cart
13-900-840
SDP
View Documents
Heidolph™
036303410
100 mL Erlenmeyer Flask
Each for $107.64
Only null left
Add to Cart
13-900-839
SDP
View Documents
Heidolph™
036303400
50mL Erlenmeyer Flask
Each for $88.99
Only null left
Add to Cart
13-900-841
SDP
View Documents
Heidolph™
036303420
250 mL Erlenmeyer Flask
Each for $123.09
Only null left
Add to Cart
13-900-842
SDP
View Documents
Heidolph™
036303430
500 mL Erlenmeyer Flask
Each for $118.45
Only null left
Add to Cart
13-900-843
SDP
View Documents
Heidolph™
036303440
1000 mL Erlenmeyer Flask
Each for $132.46
Only null left
Add to Cart
13-900-844
SDP
View Documents
Heidolph™
036303445
2000 mL Erlenmeyer Flask
Each for $186.43
Only null left
Add to Cart
Description
Description
  • Clamps provide different configurations depending on Perforated Platform and flask size used
Specifications
Specifications
Flask Clamp
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.