Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES-1 Rat anti-Human,Mouse, Unconjugated, Clone: HS395A, MilliporeSigma™
Rat Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | HES-1 |
---|---|
Clone | HS395A |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN23182MI
|
MilliporeSigma
MABN231825UL |
25 μL |
Each for $189.29
|
|
|||||
MABN23181MI
|
MilliporeSigma
MABN2318100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
HES-1 Monoclonal antibody specifically detects HES-1 in Human,Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
HES-1 | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Human, Mouse | |
Transcription factor HES-1;Hairy and enhancer of split 1 | |
His-tagged recombinant fragment corresponding to 190 amino acids from murine HES-1 protein. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
29.75 kDa calculated. |
HS395A | |
Monoclonal | |
Purified | |
Neuroscience | |
NP_032261.1 | |
Hes1, Hes-1 | |
IgG2b κ | |
Protein G Purified | |
Clone HS395A is a rat monoclonal antibody that detects HES-1 in human and murine tissues. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title