Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 125-4.6
Catalog No. p-7111330 Shop All MilliporeSigma Products
Click to view available options
Stationary Phase:
NH2
RP18
RP8
RPSELECTB
SI3
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 125-4.6 come with customized packing (please specify packing material) and are 125mmL x 4.6mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4.6 mm
Length (Metric) 125 mm
Mobile Phase Compatibility RP (when in mode) and NP
Stationary Phase NH2
Packing Material Customized (specify packing material)
Column Type Hibar™ HPLC Column
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.