Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5071984 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100479SS 0.025 mL
NB100479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100479SS Supplier Novus Biologicals Supplier No. NB100479SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 389 publications

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Canine, Fish, Goat, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.

Specifications

Antigen HIF-1 alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2ug/ml, Simple Western, Flow Cytometry 1:10 - 1:1000. Use reported in scientific literature (PMID 21917971), Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Immunohistochemistry-Frozen 5 - 10 ug/mL, Immunoblotting reported in scientific literature (PMID 28506759), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 27148974), Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q61221
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Gene Symbols HIF1A
Host Species Rabbit
Immunogen This HIF-1 alpha Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221].
Molecular Weight of Antigen 93 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091
Test Specificity Human, mouse, rat, goat, primate and rabbit. Hamster reactivity reported in scientific literature (PMID: 24997364 and 24997360).
Target Species Human, Mouse, Rat, Pig, Canine, Fish, Goat, Hamster, Primate, Rabbit
Content And Storage Store at -20°C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.