Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 13 publications
$192.94 - $517.59
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:300. Use reported in scientific literature, Immunohistochemistry-Frozen 1:100 - 1:300. Use reported in scientific literature, Gel Super Shift Assays, KnockDown Validated reported in scientific literature (PMID 32054096) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Shift, KnockDown |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100654SS
![]() |
Novus Biologicals
NB100654SS |
0.025 mL |
Each for $192.94
|
|
|||||
NB100654
![]() |
Novus Biologicals
NB100654 |
0.1 mL |
Each for $517.59
|
|
|||||
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Knockdown Validated.Specifications
| HIF-1 alpha | |
| Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:300. Use reported in scientific literature, Immunohistochemistry-Frozen 1:100 - 1:300. Use reported in scientific literature, Gel Super Shift Assays, KnockDown Validated reported in scientific literature (PMID 32054096) | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| HIF1A | |
| IgG | |
| Affinity Purified | |
| Human and porcine. Does not react with rat. Immunogen sequence has 93% homology to bovine and 90% homology to mouse.Mouse reactivity reported in scientific literature (PMID: 23959856) Rat reactivity reported in scientific literature (PMID: 24122166). |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Shift, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine | |
| Q16665 | |
| 3091 | |
| This HIF-1 alpha Antibody was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665]. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title