Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-2 alpha/EPAS1 Antibody (ep190b), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 177 publications
$244.50 - $572.50
Specifications
Antigen | HIF-2 alpha/EPAS1 |
---|---|
Clone | ep190b |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 2 ug/mL, Simple Western 1:100, Flow Cytometry 1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:150 - 1:300, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:150 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24973414), Gel Super Shift Assays reported in scientific literature (PMID 17404621), In vivo assay reported in scientific literature (PMID 23857308), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated reported in scientific literature (PMID 32054096) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100132SS
![]() |
Novus Biologicals
NB100132SS |
0.025 mL |
Each for $244.50
|
|
|||||
NB100132
![]() |
Novus Biologicals
NB100132 |
0.1 mL |
Each for $572.50
|
|
|||||
Description
HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Human, Mouse, Rat, Bovine, Hamster samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, Gel Supershift Assay, Knockdown Validated.Specifications
HIF-2 alpha/EPAS1 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown | |
Unconjugated | |
Mouse | |
Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
Q99814 | |
2034 | |
The immunogen recognized by this HIF-2 alpha/EPAS1 Antibody (ep190b) maps to a region between amino acids 535-631. [UniProt# Q99814] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
96.5 kDa |
ep190b | |
Western Blot 1 - 2 ug/mL, Simple Western 1:100, Flow Cytometry 1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:150 - 1:300, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:150 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24973414), Gel Super Shift Assays reported in scientific literature (PMID 17404621), In vivo assay reported in scientific literature (PMID 23857308), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated reported in scientific literature (PMID 32054096) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Hamster | |
Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, ep190b, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF- | |
EPAS1 | |
IgG1 | |
Protein G purified | |
This HIF-2 alpha/EPAS1 Antibody (ep190b) is specific for HIF-2 alpha/EPAS1, and does not cross-react with HIF-1 alpha. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title