Learn More
Description
Specifications
Specifications
| Antigen | HIF-2 alpha/EPAS1 |
| Applications | Flow Cytometry, KnockDown, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | ep190b |
| Conjugate | PerCP |
| Dilution | Flow Cytometry, Knockdown Validated |
| Gene Accession No. | Q99814 |
| Gene Alias | Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, |
| Gene Symbols | EPAS1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
