Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Weldment O-Rings for EZ Twist Top Split/Splitless Injection Ports
SDP

Catalog No. 06611271 Shop All Restek Corporation Products
Click to view available options
Packaging Quantity:
10/Pk.

High-Temperature, for Agilent GCs

  • High Temperature
  • For use with EZ Twist Top Split/Splitless Injection Ports

Specifications

Type Weldment O-Ring
Color Black
For Use With (Application) EZ Twist Top Split/Splitless Injection Ports
Packaging Quantity 10/Pk.
Description High-Temperature
For Use With Agilent GCs
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.