Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine Antibody, Novus Biologicals™
SDP

Catalog No. NBP24526601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
0.2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24526601 0.1 mL
NBP24526602 0.2 mL
NB413863 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24526601 Supplier Novus Biologicals Supplier No. NBP2452660.1ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Histamine in All samples, and it is validated for Dot Blot, Immunohistochemistry (Paraffin)

Histamine Polyclonal specifically detects Histamine in Human, All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot.

Specifications

Antigen Histamine
Applications Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:100 - 1:200, Dot Blot 1:14000 - 1:28000
Formulation 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide
Gene Alias 2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole
Host Species Rabbit
Immunogen Histamine conjugated to succinylated KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The product specifically stains histamine-containing cells in paraformaldehyde/carbodiimide-fixed or paraformaldehyde-fixed, paraffin-embedded sections of rat stomach (endocrine cells and mast cells).
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.