Learn More
Description
Specifically detects Histamine in All samples, and it is validated for Dot Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Histamine |
| Applications | Immunohistochemistry (Paraffin), Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:100 - 1:200, Dot Blot 1:14000 - 1:28000 |
| Formulation | 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide |
| Gene Alias | 2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole |
| Host Species | Rabbit |
| Immunogen | Histamine conjugated to succinylated KLH. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
