Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $846.50
Specifications
Antigen | Histamine |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:100 - 1:200, Dot Blot 1:14000 - 1:28000 |
Applications | Immunohistochemistry (Paraffin), Dot Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24526601
![]() |
Novus Biologicals
NBP2452660.1ML |
0.1 mL |
Each for $518.50
|
|
|||||
NBP24526602
![]() |
Novus Biologicals
NBP2452660.2ML |
0.2 mL |
Each for $846.50
|
|
|||||
NB413863
![]() |
Novus Biologicals
NBP2452660.025ML |
0.025 mL |
Each for $196.10
|
|
|||||
Description
Specifically detects Histamine in All samples, and it is validated for Dot Blot, Immunohistochemistry (Paraffin)
Histamine Polyclonal specifically detects Histamine in Human, All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot.Specifications
Histamine | |
Immunohistochemistry (Paraffin), Dot Blot | |
Unconjugated | |
RUO | |
10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
Histamine conjugated to succinylated KLH. | |
Primary | |
The product specifically stains histamine-containing cells in paraformaldehyde/carbodiimide-fixed or paraformaldehyde-fixed, paraffin-embedded sections of rat stomach (endocrine cells and mast cells). |
Immunohistochemistry-Paraffin 1:100 - 1:200, Dot Blot 1:14000 - 1:28000 | |
Polyclonal | |
Rabbit | |
All species | |
2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title