Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histidase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | Histidase |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15313520
![]() |
Novus Biologicals
NBP15313520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP153135
![]() |
Novus Biologicals
NBP153135 |
100 μL |
Each for $487.50
|
|
|||||
Description
Histidase Polyclonal specifically detects Histidase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Histidase | |
| Polyclonal | |
| Rabbit | |
| P42357 | |
| 3034 | |
| Synthetic peptides corresponding to HAL(histidine ammonia-lyase) The peptide sequence was selected from the N terminal of HAL. Peptide sequence INKLQELQVNLVRSHSSGVGKPLSPERCRMLLALRINVLAKGYSGISLET. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| EC 4.3.1, EC 4.3.1.3, HIS, Histidase, histidine ammonia-lyase, HSTD | |
| HAL | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title