Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Antibody, Novus Biologicals™
SDP

Catalog No. p-7201025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017815 0.025 mg
NB100746 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017815 Supplier Novus Biologicals Supplier No. NB1007460.025MG
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Histone H2B Polyclonal specifically detects Histone H2B in Bovine samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Histone H2B
Applications Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:1000
Formulation PBS with No Preservative
Gene Alias FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
Gene Symbols HIST2H2BF
Host Species Sheep
Immunogen Intact calf H-2B complexed to rRNA.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 440689
Test Specificity This is specific for Histone 2B.
Target Species Bovine
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.