Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3.3 Mouse anti-Human, Active Motif™

Mouse Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | Histone H3.3 |
---|---|
Concentration | 1 mg/mL |
Applications | ChIP sequencing (ChIP-seq), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3858
![]() ![]() |
Active Motif
91192 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3857
![]() ![]() |
Active Motif
91191 |
100 μg |
Each for $482.00
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.Specifications
Histone H3.3 | |
ChIP sequencing (ChIP-seq), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P84243 | |
3020 | |
This antibody was raised against a peptide corresponding to amino acids 21-39 of human Histone H3.3. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
H3-3A |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 35mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
BOS_15646; CG5825; CG5825 gene product from transcript CG5825-RA; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; H3 histone, family 3A; H3.3; H3.3A; H3.3B; H3.4; H3.A; H3.B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3B; H3FT; H3L-like histone; H3t; His3.3; His3.3A; His-3.3A; His3.3A-PA; His3.3A-PC; His3.3A-PD; His3.3B; Histone cluster 3; Histone H3; Histone H3.3; histone H3.3 variant; Histone H3.3A; Histone H3.3B; histone H3.3-like; histone variant 3.3; histone variant H3.3; LOC100054462; MGC126886; MGC126888; PH3; PP781 | |
H3-3A | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title