Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap Albumin and IgG Depletion

Catalog No. 45002171 Shop All Cytiva Products
Change view
Click to view available options
Length (Metric):
25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
45-002-171 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-171 Supplier Cytiva Supplier No. 28946603

HiTrap Albumin and IgG Depletion is a prepacked columns for the depletion of albumin and IgG from human serum and plasma. The columns are prepacked with high performance SepharoseTM based media with an affinity for human serum albumin (HSA) and IgG

  • HiTrap Albumin and IgG Depletion is a prepacked column for the depletion of albumin and IgG from human serum or plasma.
  • HiTrap Albumin and IgG Depletion column provides rapid and easy processing in a convenient format for handling larger sample volumes (∼150 μl) using a liquid chromatography system or manually using a syringe.
  • Columns are prepacked with high performance Sepharose based media with an affinity for human serum albumin (HSA) and IgG.
  • High depletion capacity, removes > 95% HSA and > 90% IgG, with high reproducibility.
  • Simple and fast procedure.
  • Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7 mm
Flow Rate <4 mL/min
Length (Metric) 25 mm
Ligand Type Recombinant Protein G fragment and recombinant Protein binding HAS
Particle Size 34 μm
pH 3 to 9
Volume (Metric) 1 mL
Endcapped No
Material Polypropylene
Product Type Guard Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.