Learn More
Description
Specifications
Specifications
| Antigen | HIV-1 Gag p24 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 7F4 |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Dilution | Western Blot 0.2 - 0.5 μg/mL, ELISA 0.2 - 0.5 μg/mL |
| Formulation | Antibody is supplied in PBS containing 1% BSA and 0.02% thimerosol. with 0.02% Thimerosal |
| Gene Alias | Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 |
| Gene Symbols | gag |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
