Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435994 0.025 mg
NBP241090 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435994 Supplier Novus Biologicals Supplier No. NBP2410900.025MG

Goat Polyclonal Antibody

HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HIV-1 gp120
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA
Formulation PBS with 0.02% Sodium Azide
Gene Alias Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein
Gene Symbols env
Host Species Goat
Immunogen Antibody was raised against 15 amino acids from near the center of gp120. The immunogen is located within amino acids 470 - 520 of gp120. Amino Acid Squence: APTKAKRRVVQREKR
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155971
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.