Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA F Antibody (3D11), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP276802A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP276802A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP276802A32 Supplier Novus Biologicals Supplier No. NBP276802AF532
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA F Monoclonal specifically detects HLA F in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen HLA F
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 3D11
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias alpha chain F, HLA class I molecule, HLA F antigen, HLA-5.4, HLAFHLA-CDA12, Leukocyte antigen F, major histocompatibility complex, class I, F, MHC class I antigen F, MHC class Ib antigen
Gene Symbols HLA-F
Host Species Mouse
Immunogen Inclusion body-derived HLA-F heavy chain
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3134
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.