Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLTF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $646.00
Specifications
Antigen | HLTF |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HLTF Polyclonal specifically detects HLTF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HLTF | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q14527 | |
6596 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LKKHGFKLGPAPKTLGFNLESGWGSGRAGPSYSMPVHAAVQMTTEQLKTEFDKLFEDLKEDDKTHEMEPAEAIETPLLPHQKQALAWMVSRENSKELPPFWEQRNDLYYNTITNFSEKDRPENVHGGILADDMGLGK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
HLTF | |
IgG | |
Affinity Purified | |
Specificity of human HLTF antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title