Learn More
Description
Specifications
Specifications
| Antigen | HNF-3 alpha/FoxA1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Classification | Monoclonal |
| Clone | FOXA1/1518 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page |
| Formulation | 10mM PBS with 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A |
| Gene Symbols | FOXA1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
