Learn More
Honeywell™ Ranger™ 1000 Boots
Offers complete protection and lightweight comfort
$383.50
Specifications
Certifications/Compliance NFPA 1971, NFPA 1992, CSA and Mustard HD Protocol TOP-8-2-501
Footbed Material Steel
For Use With (Application) Ideal to provide an insulation
Height (English) 15 in.
Height (Metric) 38.1 cm
View More Specs
Alert:
This product contains natural rubber latex proteins which may cause allergic reactions in some individuals.
Products 46
Catalog Number Mfr. No. Size Fit Price Quantity & Availability  
Catalog Number Mfr. No. Size Fit Price Quantity & Availability  
19-826-012
View Documents
Honeywell™
BT1000050N
5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-014
View Documents
Honeywell™
BT1000055N
5.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-013
View Documents
Honeywell™
BT1000050M
5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-018
View Documents
Honeywell™
BT1000065N
6.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-015
View Documents
Honeywell™
BT1000055M
5.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-017
View Documents
Honeywell™
BT1000060M
6 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-019
View Documents
Honeywell™
BT1000065M
6.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-016
View Documents
Honeywell™
BT1000060N
6 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-020
View Documents
Honeywell™
BT1000070N
7 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-021
View Documents
Honeywell™
BT1000070M
7 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-023
View Documents
Honeywell™
BT1000075M
7.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-024
View Documents
Honeywell™
BT1000080N
8 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-022
View Documents
Honeywell™
BT1000075N
7.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-025
View Documents
Honeywell™
BT1000080M
8 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-027
View Documents
Honeywell™
BT1000085M
8.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-026
View Documents
Honeywell™
BT1000085N
8.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-030
View Documents
Honeywell™
BT1000095N
9.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-029
View Documents
Honeywell™
BT1000090M
9 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-028
View Documents
Honeywell™
BT1000090N
9 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-031
View Documents
Honeywell™
BT1000095M
9.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-032
View Documents
Honeywell™
BT1000100N
10 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-033
View Documents
Honeywell™
BT1000100M
10 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-034
View Documents
Honeywell™
BT1000105N
10.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-035
View Documents
Honeywell™
BT1000105M
10.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-041
View Documents
Honeywell™
BT1000120M
12 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-036
View Documents
Honeywell™
BT1000110N
11 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-037
View Documents
Honeywell™
BT1000110M
11 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-039
View Documents
Honeywell™
BT1000115M
11.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-038
View Documents
Honeywell™
BT1000115N
11.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-040
View Documents
Honeywell™
BT1000120N
12 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-043
View Documents
Honeywell™
BT1000125M
12.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-042
View Documents
Honeywell™
BT1000125N
12.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-044
View Documents
Honeywell™
BT1000130N
13 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-047
View Documents
Honeywell™
BT1000135M
13.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-046
View Documents
Honeywell™
BT1000135N
13.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-048
View Documents
Honeywell™
BT1000140N
14 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-049
View Documents
Honeywell™
BT1000140M
14 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-045
View Documents
Honeywell™
BT1000130M
13 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-051
View Documents
Honeywell™
BT1000145M
14.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-050
View Documents
Honeywell™
BT1000145N
14.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-052
View Documents
Honeywell™
BT1000150N
15 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-057
View Documents
Honeywell™
BT1000160M
16 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-053
View Documents
Honeywell™
BT1000150M
15 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-054
View Documents
Honeywell™
BT1000155N
15.5 Narrow Width
Pair for $383.50
Only null left
Add to Cart
19-826-055
View Documents
Honeywell™
BT1000155M
15.5 Medium Width
Pair for $383.50
Only null left
Add to Cart
19-826-056
View Documents
Honeywell™
BT1000160N
16 Narrow Width
Pair for $383.50
Only null left
Add to Cart
Description
Description
A combination of natural rubber and butyl rubber ensures complete chemical protection to certified standards
  • 38cm (15 in.) Rubber Pull-on
  • Structural and liquid splash boot
  • Weight (approx.): 2.9kg (6.3 lb.)
  • Shoe-Fit™ Construction
  • Insulated
  • Steel toe, bottom plate and shank
  • Spacer Mesh Lining laminated to buoyant neoprene
  • Molded Sole with removable Insert
  • APT Barcoded
  • Made in North America
Compliance
NFPA 1971, NFPA 1992, CSA and Mustard HD Protocol TOP-8-2-501 (24 hrs)
Specifications
Specifications
NFPA 1971, NFPA 1992, CSA and Mustard HD Protocol TOP-8-2-501
Ideal to provide an insulation
38.1 cm
Steel
2.85 kg
Steel
15 in.
Natural and Butyl Rubber
6.3 lb.
No
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.