Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Hot Start Mix RTG
Shop All Cytiva Products
Click to view available options
Product Type:
illustra Hot Start Mix RTG
Quantity:
100 Reactions
480 Reactions
96 Reactions
Volume:
200μL
500μL
Description
- Primer sequestration effectively blocks greater than 99% of polymerase activity during setup effectively minimizing any nonspecific priming or primer-dimer formation
- Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
- Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment
- The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage

Specifications
Specifications
Content And Storage | 1.75 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. Ambient temperature in pouch with desiccant |
Description | Preformulated and predispensed, freeze-dried PCR reagents mix including high-quality PuReTaq DNA Polymerase and a hot start activator protein, for increased specificity and reproducibility of PCR amplifications. |
Format | Tube |
Product Type | illustra Hot Start Mix RTG |
Quantity | 96 Reactions |
For Use With (Application) | Standard PCR |
No. of Reactions | 96 |
Volume | 200μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction