Learn More
Restek™ Hot Swap Capillary Column Nuts
Shop All Restek Corporation Products
Click to view available options
For Use With (Application):
Agilent Compact Ferrules
Agilent Standard 1/16 in. Ferrules
Description
- Reduces the risk of burnt fingers
- Eliminates downtime waiting for injector parts to cool down
Specifications
Specifications
| Type | Hot Swap Capillary Column Nuts |
| Material | Stainless Steel |
| For Use With (Application) | Agilent Compact Ferrules |
| Quantity | 1 Nμt |
| For Use With (Equipment) | Agilent GCs |