Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP1 alpha Rabbit anti-Human, Mouse, Rat, Clone: 8C3M6, Novus Biologicals™
SDP

Catalog No. p-7247746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167626 20 μg
NB167625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167626 Supplier Novus Biologicals Supplier No. NBP31626720UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HP1 alpha Monoclonal antibody specifically detects HP1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen HP1 alpha
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8C3M6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:20-1:50
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 92-191 of human HP1 alpha (P45973).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 23468
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.