Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPi1 Antibody (HIC0-4F9), Novus Biologicals™
SDP

Catalog No. p-5000928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118872SS 0.025 mL
NBP118872 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP118872SS Supplier Novus Biologicals Supplier No. NBP118872SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

HPi1 Monoclonal specifically detects HPi1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen HPi1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HIC0-4F9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100. Use reported in scientific literature (PMID 29514095), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100
Formulation Tissue culture supernatant with 0.09% Sodium Azide
Gene Alias human pancreatic endocrine islet cell
Host Species Mouse
Immunogen Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells.
Purification Method Tissue culture supernatant
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Pancreatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Specific for multiple endocrine cell types.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.