Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSDL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | HSDL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17426720
![]() |
Novus Biologicals
NBP17426720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174267
![]() |
Novus Biologicals
NBP174267 |
100 μL |
Each for $487.50
|
|
|||||
Description
HSDL1 Polyclonal specifically detects HSDL1 in Human samples. It is validated for Western Blot.Specifications
HSDL1 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
hydroxysteroid dehydrogenase like 1, inactive hydroxysteroid dehydrogenase-like protein 1, MGC125994, MGC125995, MGC126032, SDR12C3, short chain dehydrogenase/reductase family 12C, member 3, steroid dehydrogenase-like protein | |
HSDL1 | |
IgG | |
37 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q3SXM5 | |
83693 | |
Synthetic peptides corresponding to the C terminal of HSDL1. Immunizing peptide sequence STLGISKRTTGYWSHSIQFLFAQYMPEWLWVWGANILNRSLRKEALSCTA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title