Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018778 0.025 mg
NBP19747505 0.05 mg
NBP19747502 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018778 Supplier Novus Biologicals Supplier No. NBP1974750.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

HSF1 Polyclonal specifically detects HSF1 in Human, Mouse, Rat, Gerbil, Primate, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Gel Super Shift Assays, Enzyme Immunoassay.

Specifications

Antigen HSF1
Applications ELISA, Western Blot, Gel Shift, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. Q00613
Gene Alias heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Gene Symbols HSF1
Host Species Rabbit
Immunogen Recombinant human HSF1.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3297
Test Specificity Recognizes nonphosphorylated HSF1 at ∼60kDa and phosphorylated HSF1 at ∼80kDa.
Target Species Primate, Rat, Rabbit, Human, Mouse, Gerbil
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.