Learn More
Promega HSM 2.0 Instrument, 1 each
Catalog No. PRA2715 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRA2715 1 Ea.
1 options
Catalog No. PRA2715 Supplier Promega Supplier No. A2715
Add to Cart
Add to Cart
Request a Quote
The Heater Shaker Magnet Instrument (HSM 2.0) performs all the functions necessary for the processing of magnetic resin-based purification chemistries in large-volume formats.
  • All−In−One Processing for Large−Volume Purification Chemistries
  • Processes magnetic resin−based purification chemistries in large−volume formats
  • Uses standard 50ml conical tubes, magnets and reagent−based paramagnetic particles
  • Preprogrammed with methods for processing up to 32 whole blood samples
Specifications
Description 1 each
Quantity 1 Ea.