Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP90 Antibody (2D12), Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications
$243.50 - $719.00
Specifications
Antigen | HSP90 |
---|---|
Clone | 2D12 |
Dilution | Western Blot 1:500, Immunoprecipitation 1:10-1:500 |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018766
![]() |
Novus Biologicals
NBP1974500.025ML |
0.025 mL |
Each for $243.50
|
|
|||||
NBP19745005
![]() |
Novus Biologicals
NBP1974500.05ML |
0.05 mL |
Each for $402.00
|
|
|||||
NBP19745002
![]() |
Novus Biologicals
NBP1974500.2ML |
0.2 mL |
Each for $719.00
|
|
|||||
Description
HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Fungi, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HSP90 | |
Western Blot 1:500, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
3320 | |
Native mouse Hsp90. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
2D12 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Hamster, Rat, Rabbit, Primate, Chicken, Sheep, Mouse, Fish, Bovine, Guinea Pig, Human, Fungi, Canine | |
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
HSP90AA1 | |
IgM | |
Ammonium sulfate precipitation | |
Reacts with Beluga, and water mold |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title