Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HT-29 Whole Cell Lysate
SDP

Catalog No. nbp20396301 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.1 mg

Applications: Western Blot

The HT-29 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Tissue HT-29
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity HT-29 (Human Colorectal Adenocarcinoma)
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.