Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTRA2/Omi Antibody (196C429), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen HTRA2/Omi
Clone 196C429
Dilution Western Blot 2-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (Lee et al (2003))
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056558T
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NB10056558SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056558
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NB10056558
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

HTRA2/Omi
Western Blot 2-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (Lee et al (2003))
Monoclonal
Purified
RUO
Human
High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI
HTRA2
IgG1 κ
Protein G purified
This antibody does not cross-react with mouse HtrA2.
196C429
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cancer, Mitochondrial Mediated Pathway
NP_713130.1
27429
This monoclonal antibody was developed by immunizing mice with a synthetic peptide containing amino acids 335-350 of human HtrA2.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.