Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTRA2/Omi Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | HTRA2/Omi |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HTRA2/Omi Polyclonal specifically detects HTRA2/Omi in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HTRA2/Omi | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, Mitochondrial Mediated Pathway | |
High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
HTRA2 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
27429 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RLREFLHRGEKKNSSSGISGSQRRYIGVMMLTLSPSILAELQLREPSFPDVQHGVLIHKVILGSP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title