Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-1/CD80 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
$227.50 - $2070.00
Specifications
Antigen | B7-1/CD80 |
---|---|
Clone | 37711 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 0.2-1 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF, In Situ Hybridization (ISH), ELISA |
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready, Dual RNAscope ISH-IHC, ELISA Capture (Matched Antibody Pair).Specifications
B7-1/CD80 | |
LYOPH | |
Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF, In Situ Hybridization (ISH), ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
CD80 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
37711 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 0.2-1 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P33681 | |
941 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-1/CD80 Val35-Asn242 (predicted) Accession # P33681 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human B7-1/CD80 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) B7-2, recombinant mouse (rm) B7-1, rmB7-2, rhB7-H1, rhB7-H2, rhB7-H3 or rmPD-L2 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title