Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Phospho- Histone H2AX (S139) Antibody, R&D Systems™

Rabbit Antibody

Specifications

Antigen Histone H2AX
Dilution Western Blot 1:2500 dilution, Immunocytochemistry 1.7-15 ug/mL
Conjugate Unconjugated
Target Species Human, Mouse
Formulation Supplied as a solution containing PBS.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
44-18A-PC020
View Documents
R&D Systems
4418APC020
20 μL N/A N/A N/A
44-18A-PC100
View Documents
R&D Systems
4418APC100
100 μL N/A N/A N/A
Description

Description

Histone H2AX specifically detects Histone H2AX in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

Histone H2AX
Unconjugated
Supplied as a solution containing PBS.
3014
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening.
Western Blot 1:2500 dilution, Immunocytochemistry 1.7-15 ug/mL
Human, Mouse
H2A.X, H2A/X, H2AFX
Phosphopeptide containing human Histone H2AX S139 site
Antigen Affinity-purified
Detects human and mouse Histone H2AX when phosphorylated at S139 in Western blots. This antibody detects only double-strand DNA breaks.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.