Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Human Papilloma Virus (HPV) Ab-3, Mouse Monoclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$513.00 - $1090.00

Specifications

Antigen Human Papilloma Virus (HPV) Ab-3
Clone K1H8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1826S1
View Documents
Epredia™
MS1826S1A
500 μL
Each for $602.00
Only null left
Add to Cart
 
MS1826S
View Documents
Epredia™
MS1826SA
1 mL
Each for $1,090.00
Only null left
Add to Cart
 
MS1826R7
View Documents
Epredia™
MS1826R7A
7 mL
Each for $513.00
Only null left
Add to Cart
 
MS1826S0
View Documents
Epredia™
MS1826S0A
100 μL N/A N/A N/A
MS1826RQ
View Documents
Epredia™
MS1826RQA
12 mL N/A N/A N/A
Description

Description

Human papilloma virus is non-enveloped double stranded circular DNA virus with icosahedral capsid.

Host Species: Mouse

Clone: K1H8

Isotype: IgG1/κ

Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.

Epitope: Nonconfirmational internal linear epitope of major capsid protein

Immunogen: Alkaline disrupted HPV type 1

Molecular Weight: 57kDa (major band)

Positive Control: Infected cells and tissue

Cellular Localization: Nuclei of infected cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Human Papilloma Virus (HPV) Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
ASR, IVD
Alkaline disrupted HPV type 1
K1H8
Monoclonal
Mouse
Infectious Disease
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.