Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SIT1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | SIT1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
SIT1 Polyclonal specifically detects SIT1 in Human samples. It is validated for Western Blot.Specifications
SIT1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1 | |
SIT1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9Y3P8 | |
27240 | |
E. coli-derived recombinant human SIT1 Gln65-Ser196 Accession # Q9Y3P8 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human SIT1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title