Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HUS1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | HUS1B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15458220
![]() |
Novus Biologicals
NBP15458220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154582
![]() |
Novus Biologicals
NBP154582 |
100 μL |
Each for $487.50
|
|
|||||
Description
HUS1B Polyclonal specifically detects HUS1B in Human samples. It is validated for Western Blot.Specifications
HUS1B | |
Polyclonal | |
Rabbit | |
Human | |
checkpoint protein HUS1B, hHUS1B, HUS1 (S. pombe) checkpoint homolog b, HUS1 checkpoint homolog b (S. pombe), MGC126746, MGC126748, RP11-532F6.1 | |
HUS1B | |
IgG | |
31 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q8NHY5 | |
135458 | |
Synthetic peptides corresponding to HUS1B(HUS1 checkpoint homolog b (S. pombe)) The peptide sequence was selected from the N terminal of HUS1B. Peptide sequence ELFIHVSGTVARLAKVCVLRVRPDSLCFGPAGSGGLHEARLWCEVRQGAF. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title