Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS Antibody (2317C), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349735 Supplier Novus Biologicals Supplier No. FAB1682MFV500
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ICOS Monoclonal antibody specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry, Blocking Assay, CyTOF

Specifications

Antigen ICOS
Applications Flow Cytometry, Blocking Assay, CyTOF
Classification Monoclonal
Clone 2317C
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived mouse ICOS, Met1-Leu142, Accession # Q9WVS0
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 29851
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.