Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha Antibody (967370), R&D Systems™
SDP

Catalog No. p-7078957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB93451SP 25 μg
MAB93451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB93451SP Supplier Novus Biologicals Supplier No. MAB93451SP
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for Block/Neutralize.

Specifications

Antigen IFN-alpha
Applications Blocking Assay
Classification Monoclonal
Clone 967370
Conjugate Unconjugated
Dilution Blocking/Neutralizing
Formulation PBS with No Preservative
Gene Alias IFNA, IFNA2, IFNA2b, IFN-alphaA, INFA2, interferon alpha 2
Gene Symbols IFNA2
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha beta R1 Lys28-Lys436 Accession # AAA52730
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3440
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.