Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHMBP2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen IGHMBP2
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB16901120
SDP
View Documents
Novus Biologicals
NBP16892120UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP168921
SDP
View Documents
Novus Biologicals
NBP168921
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

IGHMBP2 Polyclonal specifically detects IGHMBP2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

IGHMBP2
Unconjugated
RUO
ATP-dependent helicase IGHMBP2, CATF1FLJ41171, EC 3.6.1, EC 3.6.4.12, EC 3.6.4.13, GF-1, Glial factor 1, HCSA, HMN6cardiac transcription factor 1, immunoglobulin mu binding protein 2, Immunoglobulin mu-binding protein 2, SMARD1DNA-binding protein SMUBP-2, SMBP2, SMUBP2FLJ34220
IGHMBP2
Primary
Polyclonal
Rabbit
P40694
3508
IgG
109 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.