Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF2/CD101 Antibody (BB27) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017699 0.025 mg
NB10065269 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017699 Supplier Novus Biologicals Supplier No. NB100652690.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGSF2/CD101 Monoclonal specifically detects IGSF2/CD101 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen IGSF2/CD101
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunohistochemistry
Classification Monoclonal
Clone BB27
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Accession No. Q93033
Gene Alias CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2
Gene Symbols CD101
Host Species Mouse
Immunogen Human thymic clone B12
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9398
Test Specificity NB100-65269 recognizes human CD101. CD101 is a 140kD homodimeric cell surface glycoprotein expressed primarily by monocytes, granulocytes, dendritic cells and activated T lymphocytes. Studies suggest that CD101 plays a major role in the activation of T cells by skin dendritic cells. Clone BB27 has been reported to inhibit allogeneic T-cell responses (1). Removal of sodium azide is recommended prior to use in functional assays.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.