Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF8/CD316 Antibody (8A12), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB260425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260425 Supplier Novus Biologicals Supplier No. NBP321498AF405
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGSF8/CD316 Monoclonal antibody specifically detects IGSF8/CD316 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen IGSF8/CD316
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 8A12
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8
Host Species Mouse
Immunogen Human leukocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 93185
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.