Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IIGP1 Antibody, Novus Biologicals™
SDP

Catalog No. p-5066264 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203970SS 0.025 mg
NBP203970 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP203970SS Supplier Novus Biologicals Supplier No. NBP203970SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IIGP1 Polyclonal specifically detects IIGP1 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen IIGP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_068564.4
Gene Alias 2900074L10Rik, AI046432, AW111922, Ifgga1, Iigp, Iigp1, Irga6
Gene Symbols Iigp1
Host Species Rabbit
Immunogen A portion of amino acids 325-375 of mouse Iigp1 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60440
Test Specificity This antibody will detect both two isoforms of Iigp1 (413 amino acids and 346 amino acids).
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.