Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody (MM0070-7D24) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
|---|---|
| Clone | MM0070-7D24 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot.Specifications
| 2B4/CD244/SLAMF4 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| CD244 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0070-7D24 | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_057466 | |
| 51744 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title